Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 14 (MMP14) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05842
Encompass_Preferred
Mouse Matrix Metalloproteinase 14 (MMP14) ELISA Kit for detecting MMP14 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53690. Gene ID: 17387. Target Synonyms: MMP-X1; MT1-MMP; MTMMP1; Membrane Inserted; Membrane-type matrix metalloproteinase 1; Membrane-type-1 matrix metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3226