Learn More
Biomatik Corporation Human SHANK3 (SH3 and multiple ankyrin repeat domains protein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF60824
Encompass_Preferred
Human SHANK3 (SH3 and multiple ankyrin repeat domains protein 3) ELISA Kit for detecting SHANK3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9BYB0. Target Synonyms: SH3 and multiple ankyrin repeat domains protein 3; Shank3; Proline-rich synapse-associated protein 2; ProSAP2; KIAA1650; PROSAP2; PSAP2; SHANK3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8480