Learn More
Biomatik Corporation Human Matrix Metalloproteinase 2 (MMP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05850
Encompass_Preferred
The Human Matrix Metalloproteinase 2 (MMP2) ELISA Kit allows for quantitative determination of Matrix Metalloproteinase 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08253. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3230