Learn More
Biomatik Corporation FLAG-Tag (DYKDDDDK-Tag Protein) ELISA Kit
Catalog No. 502288484
Encompass_Preferred
FLAG-Tag (DYKDDDDK-Tag Protein) ELISA Kit for detecting FLAG-Tag in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 15.625-1000ng/ml. Sensitivity: 9.375ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: DYKDDDDK. Target Synonyms: FLAG-Tag; DYKDDDDK-Tag Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-228-8484 Supplier Biomatik Corporation Supplier No. EKF60835
FLAG-Tag (DYKDDDDK-Tag Protein) ELISA Kit for detecting FLAG-Tag in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 15.625-1000ng/ml. Sensitivity: 9.375ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: DYKDDDDK. Target Synonyms: FLAG-Tag; DYKDDDDK-Tag Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.