Learn More
Biomatik Corporation 8-OHdG (8-Hydroxydeoxyguanosine) ELISA Kit
Supplier:  Biomatik Corporation EKF60843
Encompass_Preferred
8-OHdG (8-Hydroxydeoxyguanosine) ELISA Kit for detecting 8-OHdG in serum, plasma, cell culture supernatant, cell or tissue lysate, urine, other biological fluids. Species Reactivity: Universal. Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: 8-OHdG(8-Hydroxydeoxyguanosine)/8-oxo-DG/ 8-hydroxy一2 deoxyguanosine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8489