Learn More
Biomatik Corporation Pig Matrix Metalloproteinase 3 (MMP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05865
Encompass_Preferred
Pig Matrix Metalloproteinase 3 (MMP3) ELISA Kit for detecting MMP3 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 4.69-300ng/ml. Sensitivity: 1.8ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1SV58. Target Synonyms: SL1; STMY; STMY1; STR1; Progelatinase; Stromelysin 1; Transin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3233