Learn More
Biomatik Corporation Mouse ADP/Acrp30 (Adiponectin) ELISA Kit
Supplier:  Biomatik Corporation EKE61474
Encompass_Preferred
Mouse ADP/Acrp30 (Adiponectin) ELISA Kit for detecting ADP, Acrp30 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q60994. Target Synonyms: Acrp30; GBP28; ACDC; APM1; ADPN; AdipoQ; ADIPQTL1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8490