Learn More
Biomatik Corporation Cattle Matrix Metalloproteinase 7 (MMP7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05868
Encompass_Preferred
The Cattle Matrix Metalloproteinase 7 (MMP7) ELISA Kit allows for quantitative determination of Matrix Metalloproteinase 7 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q148N3. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 7 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3235