Learn More
Biomatik Corporation Mouse IL-10 (Interleukin 10) ELISA Kit
Supplier:  Biomatik Corporation EKE61493
Encompass_Preferred
Mouse IL-10 (Interleukin 10) ELISA Kit for detecting IL-10 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.63-1000pg/ml. Sensitivity: 9.38pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P18893. Target Synonyms: IL10; IL10A; CSIF; TGIF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8495