Learn More
Biomatik Corporation Horse Matrix Metalloproteinase 9 (MMP9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05880
Encompass_Preferred
The Horse Matrix Metalloproteinase 9 (MMP9) ELISA Kit allows for quantitative determination of Matrix Metalloproteinase 9 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Equus caballus; Equine (Horse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: A8HDR2. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 9 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3240