Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 9 (MMP9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05882
Encompass_Preferred
Mouse Matrix Metalloproteinase 9 (MMP9) ELISA Kit for detecting MMP9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41245. Gene ID: 17395. Target Synonyms: GELB; Gelatinase B; CLG4B; CLG4-B; 92 KDa Gelatinase; 92kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3242