Learn More
Biomatik Corporation Human Mdm2 p53 Binding Protein Homolog (MDM2) ELISA Kit, 96 tests
Catalog No. 501503245
Encompass_Preferred
The Human Mdm2 p53 Binding Protein Homolog (MDM2) ELISA Kit allows for quantitative determination of Mdm2 p53 Binding Protein Homolog in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q00987. Cross Activity: No significant cross-reactivity or interference between Mdm2 p53 Binding Protein Homolog and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3245 Supplier Biomatik Corporation Supplier No. EKU05887
The Human Mdm2 p53 Binding Protein Homolog (MDM2) ELISA Kit allows for quantitative determination of Mdm2 p53 Binding Protein Homolog in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q00987. Cross Activity: No significant cross-reactivity or interference between Mdm2 p53 Binding Protein Homolog and analogues was observed. Restriction: For Research Use Only.