Learn More
Biomatik Corporation Mouse ACTα2 (Actin Alpha 2, Smooth Muscle) ELISA Kit
Supplier:  Biomatik Corporation EKE61867
Encompass_Preferred
Mouse ACTα2 (Actin Alpha 2, Smooth Muscle) ELISA Kit for detecting ACTα2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.13-200ng/ml. Sensitivity: 1.88ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P62737. Target Synonyms: ACTA2; AAT6; ACTSA; MYMY5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8520