Learn More
Biomatik Corporation Monkey TTR (Transthyretin) ELISA Kit
Supplier:  Biomatik Corporation EKE61948
Encompass_Preferred
Monkey TTR (Transthyretin) ELISA Kit for detecting TTR in serum, plasma, other biological fluids. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.31-20ug/ml. Sensitivity: 0.19ug/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8522