Learn More
Biomatik Corporation Mouse Melanoma Cell Adhesion Molecule (MCAM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05894
Encompass_Preferred
Mouse Cluster Of Differentiation 146 (CD146) ELISA Kit for detecting CD146 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.253ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R2Y2. Gene ID: 84004. Target Synonyms: MCAM; MUC18; Melanoma Cell Adhesion Molecule; Cell surface glycoprotein P1H12; Melanoma-associated antigen A32; S-endo 1 endothelial-associated antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3249