Learn More
Biomatik Corporation Melatonin (MT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05900
Encompass_Preferred
Melatonin (MT) ELISA Kit for detecting MT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.63pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: N-Acetyl-5-Methoxytryptamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3250