Learn More
Biomatik Corporation Human Membrane Spanning 4 Domains Subfamily A, Member 1 (MS4A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05906
Encompass_Preferred
The Human Membrane Spanning 4 Domains Subfamily A, Member 1 (MS4A1) ELISA Kit allows for quantitative determination of Membrane Spanning 4 Domains Subfamily A, Member 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.25-80 ng/mL. Sensitivity: 0.57 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11836. Cross Activity: No significant cross-reactivity or interference between Membrane Spanning 4 Domains Subfamily A, Member 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3253