Learn More
Biomatik Corporation Human Mesencephalic Astrocyte Derived Neurotrophic Factor (MANF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05912
Encompass_Preferred
The Human Mesencephalic Astrocyte Derived Neurotrophic Factor (MANF) ELISA Kit allows for quantitative determination of Mesencephalic Astrocyte Derived Neurotrophic Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55145. Cross Activity: No significant cross-reactivity or interference between Mesencephalic Astrocyte Derived Neurotrophic Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3255