Learn More
Biomatik Corporation Mouse Metallothionein 1 (MT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05917
Encompass_Preferred
Mouse Metallothionein 1 (MT1) ELISA Kit for detecting MT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02802. Gene ID: 17748. Target Synonyms: MT1A; MT1S; MTC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3258