Learn More
Biomatik Corporation Rat Metallothionein 1 (MT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05918
Encompass_Preferred
Rat Metallothionein 1 (MT1) ELISA Kit for detecting MT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 14.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02803. Gene ID: 24567. Target Synonyms: MT1A; MT1S; MTC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3259