Learn More
Biomatik Corporation Rat BDNF (Brain Derived Neurotrophic Factor) ELISA Kit
Supplier:  Biomatik Corporation EKE62350
Encompass_Preferred
Rat BDNF (Brain Derived Neurotrophic Factor) ELISA Kit for detecting BDNF in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P23363. Target Synonyms: ANON2; BULN2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8560