Learn More
Biomatik Corporation Human Microtubule Associated Protein 2 (MAP2) ELISA Kit, 96 tests
Catalog No. 501503265
Encompass_Preferred
The Human Microtubule Associated Protein 2 (MAP2) ELISA Kit allows for quantitative determination of Microtubule Associated Protein 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11137. Cross Activity: No significant cross-reactivity or interference between Microtubule Associated Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3265 Supplier Biomatik Corporation Supplier No. EKU05950
The Human Microtubule Associated Protein 2 (MAP2) ELISA Kit allows for quantitative determination of Microtubule Associated Protein 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11137. Cross Activity: No significant cross-reactivity or interference between Microtubule Associated Protein 2 and analogues was observed. Restriction: For Research Use Only.