Learn More
Biomatik Corporation 5-Hydroxytryptamine (5-HT) ELISA Kit
Supplier:  Biomatik Corporation EKL55118
Encompass_Preferred
5-Hydroxytryptamine (5-HT) ELISA Kit for detecting 5-HT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 15.6-1000ng/ml. Sensitivity: 4.16ng/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 3h. Target Synonyms: 5HT; Serotonin; N-Methyl-Gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8617