Learn More
Biomatik Corporation Mouse Microtubule Associated Protein Tau (MAPt) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05956
Encompass_Preferred
Mouse Tau Protein (MAPT) ELISA Kit for detecting MAPT in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10637. Gene ID: 17762. Target Synonyms: DDPAC; FTDP-17; MAPTL; MSTD; MTBT1; MTBT2; PPND; Neurofibrillary tangle protein; Microtubule Associated Protein Tau; Paired helical filament-tau; G Protein Beta1/Gamma2 Subunit-Interacting Factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3267