Learn More
Biomatik Corporation Human Adrenergic Receptor Beta 1 (ADRb1) ELISA Kit
Supplier:  Biomatik Corporation EKL55297
Encompass_Preferred
Human Adrenergic Receptor Beta 1 (ADRb1) ELISA Kit for detecting ADRb1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Target Synonyms: ADR-B1; ADRB1R; B1AR; BETA1AR; RHR; Beta 1 Adrenoreceptor; Beta-1 adrenoceptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8630