Learn More
Biomatik Corporation Mouse Midkine (MK) ELISA Kit, 96 tests
Catalog No. 501503269
Encompass_Preferred
Mouse Midkine (MK) ELISA Kit for detecting MK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12025. Gene ID: 17242. Target Synonyms: MDK; ARAP; NEGF2; Neurite Growth-Promoting Factor 2; Amphiregulin-associated protein; Midgestation and kidney protein; Neurite outgrowth-promoting factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3269 Supplier Biomatik Corporation Supplier No. EKU05960
Mouse Midkine (MK) ELISA Kit for detecting MK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12025. Gene ID: 17242. Target Synonyms: MDK; ARAP; NEGF2; Neurite Growth-Promoting Factor 2; Amphiregulin-associated protein; Midgestation and kidney protein; Neurite outgrowth-promoting factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.