Learn More
Biomatik Corporation Human Milk Fat Globule EGF Factor 8 (MFGE8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05963
Encompass_Preferred
The Human Milk Fat Globule EGF Factor 8 (MFGE8) ELISA Kit allows for quantitative determination of Milk Fat Globule EGF Factor 8 in Serum, Plasma, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q08431. Cross Activity: No significant cross-reactivity or interference between Milk Fat Globule EGF Factor 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3270