Learn More
Biomatik Corporation Human Gastrin (GT) ELISA Kit
Supplier:  Biomatik Corporation EKL57736
Encompass_Preferred
Human Gastrin (GT) ELISA Kit for detecting GT in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 4.36pg/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 3h. Accession Number: P01350. Gene ID: 2520. Target Synonyms: GAST; GAS; Gastrin component I; Big gastrin; Gastrin component II; Gastrin component III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8699