Learn More
Biomatik Corporation Simian Monocyte Chemotactic Protein 1 (MCP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05989
Encompass_Preferred
Simian Monocyte Chemotactic Protein 1 (MCP1) ELISA Kit for detecting MCP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61275. Gene ID: 574138. Target Synonyms: CCL2; GDCF2; HC11; HSMCR30; MCAF; MCP1; SCYA2; SMC-CF; Chemokine C-C-Motif Ligand 2; Monocyte Chemotactic And Activating Factor; Monocyte Secretory Protein JE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3277