Learn More
Biomatik Corporation Human Monocyte Chemotactic Protein 3 (MCP3) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU05993
The Human Monocyte Chemotactic Protein 3 (MCP3) ELISA Kit allows for quantitative determination of Monocyte Chemotactic Protein 3 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P80098. Cross Activity: No significant cross-reactivity or interference between Monocyte Chemotactic Protein 3 and analogues was observed. Restriction: For Research Use Only.