Learn More
Biomatik Corporation Rat Monocyte Chemotactic Protein 3 (MCP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05995
Encompass_Preferred
Rat Monocyte Chemotactic Protein 3 (MCP3) ELISA Kit for detecting MCP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QXY8. Gene ID: 287561. Target Synonyms: CCL7; FIC; MARC; NC28; MCP3; SCYA7; Chemokine C-C-Motif Ligand 7; Small Inducible Cytokine A7; Monocyte Chemoattractant Protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3280