Learn More
Biomatik Corporation Human carbonhydrate antigen 19-9 (CA19-9) ELISA kit
Supplier:  Biomatik Corporation EKC42224
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 10 U/mL-150 U/mL. Sensitivity: 3.8 U/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: CA199. Restriction: For Research Use Only.
Catalog No. 50-228-7854