Learn More
Biomatik Corporation Mouse Monocyte Chemotactic Protein 5 (MCP5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05996
Encompass_Preferred
Mouse Monocyte Chemotactic Protein 5 (MCP5) ELISA Kit for detecting MCP5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62401. Gene ID: 20293. Target Synonyms: CCL12; SCYA12; Chemokine C-C-Motif Ligand 12; MCP-1-related chemokine; Monocyte chemoattractant protein 5; Small-inducible cytokine A12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3281