Learn More
Biomatik Corporation Human Adp-Ribosylation Factor-Like Protein 8B (Arl8B) Elisa Kit
Supplier:  Biomatik Corporation EKC42377
Encompass_Preferred
Human Adp-Ribosylation Factor-Like Protein 8B (Arl8B) Elisa Kit for detecting Adp-Ribosylation Factor-Like 8B in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NVJ2; ARL8B. Target Synonyms: ADP ribosylation factor like 10C; ADP ribosylation factor like 8B; ADP-ribosylation factor-like protein 10C; ADP-ribosylation factor-like protein 8B; ARL10C; ARL8B; ARL8B_HUMAN; FLJ10702; Gie1; Novel small G protein indispensable for equal chromosome segregation 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7880