Learn More
Biomatik Corporation Human Mucin 2 (MUC2) ELISA Kit, 96 tests
Catalog No. 501503289
Encompass_Preferred
The Human Mucin 2 (MUC2) ELISA Kit allows for quantitative determination of Mucin 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.57 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q02817. Cross Activity: No significant cross-reactivity or interference between Mucin 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3289 Supplier Biomatik Corporation Supplier No. EKU06020
The Human Mucin 2 (MUC2) ELISA Kit allows for quantitative determination of Mucin 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.57 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q02817. Cross Activity: No significant cross-reactivity or interference between Mucin 2 and analogues was observed. Restriction: For Research Use Only.