Learn More
Biomatik Corporation Human SDF-1 (Stromal Cell Derived Factor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57709
Encompass_Preferred
Human SDF-1 (Stromal Cell Derived Factor 1) ELISA Kit for detecting SDF-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48061. Target Synonyms: SDF-1; CXCL12; Stromal Cell Derived Factor 1; PBSF; hIRH; hSDF-1; SCYB12; SDF1A; SDF1B; SDF1; SDF-1b; TLSF; TLSF-a; TLSF-b; IRH; SDF-1a; TPAR1; chemokine; C-X-C motif ligand 12; C-X-C motif chemokine 12; Intercrine reduced in hepatomas; Pre-B cell growth-stimulating factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7991