Learn More
Biomatik Corporation Human BDG (beta-D-glucan) ELISA Kit
Supplier:  Biomatik Corporation EKF58058
Encompass_Preferred
Human BDG (beta-D-glucan) ELISA Kit for detecting BDG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Target Synonyms: beta-D-glucan. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8028