Learn More
Biomatik Corporation Human IgA2 (Immumoglobulin A2) ELISA Kit
Supplier:  Biomatik Corporation EKF58133
Encompass_Preferred
Human IgA2 (Immumoglobulin A2) ELISA Kit for detecting IgA2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: HRP Antibody, Sandwich ELISA. Assay Time: 3h, 30 min. Accession Number: P01877. Target Synonyms: IgA2; Immumoglobulin A2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8041