Learn More
Biomatik Corporation Mouse NLRP3 (Nod Like Receptor Pyrins-3) ELISA Kit
Supplier:  Biomatik Corporation EKF58329
Encompass_Preferred
Mouse NLRP3 (Nod Like Receptor Pyrins-3) ELISA Kit for detecting NLRP3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8R4B8. Target Synonyms: Nod Like Receptor Pyrins-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8097