Learn More
Biomatik Corporation Human Myelin Associated Glycoprotein (MAG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06048
Encompass_Preferred
The Human Myelin Associated Glycoprotein (MAG) ELISA Kit allows for quantitative determination of Myelin Associated Glycoprotein in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P20916. Cross Activity: No significant cross-reactivity or interference between Myelin Associated Glycoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3299