Learn More
Biomatik Corporation Human Myelin Basic Protein (MBP) ELISA Kit, 96 tests
Catalog No. 501503300
Encompass_Preferred
The Human Myelin Basic Protein (MBP) ELISA Kit allows for quantitative determination of Myelin Basic Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02686. Cross Activity: No significant cross-reactivity or interference between Myelin Basic Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3300 Supplier Biomatik Corporation Supplier No. EKU06050
The Human Myelin Basic Protein (MBP) ELISA Kit allows for quantitative determination of Myelin Basic Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02686. Cross Activity: No significant cross-reactivity or interference between Myelin Basic Protein and analogues was observed. Restriction: For Research Use Only.