Learn More
Biomatik Corporation Human TLR4 (Toll-like receptor 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58579
Encompass_Preferred
Human TLR4 (Toll-like receptor 4) ELISA Kit for detecting TLR4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00206. Target Synonyms: TLR4; CD284; ARMD10; CD284 antigen; hTollhomolog of Drosophila toll; TOLL; toll-like receptor 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8154