Learn More
Biomatik Corporation Mouse Myelin Basic Protein (MBP) ELISA Kit, 96 tests
Catalog No. 501503301
Encompass_Preferred
Mouse Myelin Basic Protein (MBP) ELISA Kit for detecting MBP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04370. Gene ID: 17196. Target Synonyms: Myelin A1 protein; Myelin membrane encephalitogenic protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3301 Supplier Biomatik Corporation Supplier No. EKU06051
Mouse Myelin Basic Protein (MBP) ELISA Kit for detecting MBP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04370. Gene ID: 17196. Target Synonyms: Myelin A1 protein; Myelin membrane encephalitogenic protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.