Learn More
Biomatik Corporation Rat Myelin Basic Protein (MBP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06052
Encompass_Preferred
Rat Myelin Basic Protein (MBP) ELISA Kit for detecting MBP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02688. Gene ID: 24547. Target Synonyms: Myelin A1 protein; Myelin membrane encephalitogenic protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3302