Learn More
Biomatik Corporation Human CYCS (Cytochrome c) ELISA Kit
Supplier:  Biomatik Corporation EKF58832
Encompass_Preferred
Human CYCS (Cytochrome c) ELISA Kit for detecting CYCS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P99999. Target Synonyms: CYCS; Cytochrome c; CYCHCS; cytochrome c; somatic; THC4; CytC; CYC; HCS; THC4; Cyt-C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8191