Learn More
Biomatik Corporation Human Myeloid Progenitor Inhibitory Factor 1 (MPIF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06062
Encompass_Preferred
The Human Myeloid Progenitor Inhibitory Factor 1 (MPIF1) ELISA Kit allows for quantitative determination of Myeloid Progenitor Inhibitory Factor 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.62-1000 pg/mL. Sensitivity: 0.47 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55773. Cross Activity: No significant cross-reactivity or interference between Myeloid Progenitor Inhibitory Factor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3304