Learn More
Biomatik Corporation Human anti-EPO antibody (anti-Erythropoietin antibody) ELISA Kit
Supplier:  Biomatik Corporation EKF58886
Encompass_Preferred
Human anti-EPO antibody (anti-Erythropoietin antibody) ELISA Kit for detecting anti-EPO antibody in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antigen. Assay Time: 3h, 30 min. Accession Number: P01588. Target Synonyms: anti-EPO antibody; anti-Erythropoietin antibody. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8200