Learn More
Biomatik Corporation Human G6PD (Glucose-6-phosphate 1-dehydrogenase) ELISA Kit
Supplier:  Biomatik Corporation EKF58905
Encompass_Preferred
Human G6PD (Glucose-6-phosphate 1-dehydrogenase) ELISA Kit for detecting G6PD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11413. Target Synonyms: G6PD; Glucose-6-phosphate 1-dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8206