Learn More
Biomatik Corporation Mouse Myeloperoxidase (MPO) ELISA Kit, 96 tests
Catalog No. 501503307
Encompass_Preferred
Mouse Myeloperoxidase (MPO) ELISA Kit for detecting MPO in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11247. Gene ID: 17523. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3307 Supplier Biomatik Corporation Supplier No. EKU06068
Mouse Myeloperoxidase (MPO) ELISA Kit for detecting MPO in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11247. Gene ID: 17523. Restrictions: For Research Use Only. Not for use in diagnostic procedures.