Learn More
Biomatik Corporation Pig Myeloperoxidase (MPO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06069
Encompass_Preferred
Pig Myeloperoxidase (MPO) ELISA Kit for detecting MPO in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 78-5,000pg/ml. Sensitivity: 35pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1RRP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3308